missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylethynylphthalic Anhydride 98.0+%, TCI America™

Catalog No. P14375G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14375G 5 g
P143725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14375G Supplier TCI America Supplier No. P14375G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 119389-05-8
Molecular Formula C16H8O3
Molecular Weight (g/mol) 248.237
MDL Number MFCD01862632
InChI Key UPGRRPUXXWPEMV-UHFFFAOYSA-N
PubChem CID 11053989
IUPAC Name 5-(2-phenylethynyl)-2-benzofuran-1,3-dione
SMILES C1=CC=C(C=C1)C#CC2=CC3=C(C=C2)C(=O)OC3=O

Specifications

Melting Point 152°C
Color Yellow
Quantity 5 g
Formula Weight 248.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylethynylphthalic Anhydride