missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylbutyric acid, 99%

Catalog No. AAA1811514
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1811514 25 g
AAA1811522 100 g
AAA1811536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1811514 Supplier Thermo Scientific Chemicals Supplier No. A1811514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1821-12-1 | C10H12O2 | 164.204 g/mol

4-Phenylbutyric acid is used as a chemical chaperone involved in protein-folding disorders. It is involved in the synthesis of dyes and an active pharmaceutical ingredient intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenylbutyric acid is used as a chemical chaperone involved in protein-folding disorders. It is involved in the synthesis of dyes and an active pharmaceutical ingredient intermediate.

Solubility
Soluble in water and methanol.

Notes
Styrene-DVB; Strongly basic gel type resin.Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1821-12-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00004403
InChI Key OBKXEAXTFZPCHS-UHFFFAOYSA-N
Synonym 4-phenylbutyric acid, benzenebutanoic acid, benzenebutyric acid, phenylbutyrate, gamma-phenylbutyric acid, omega-phenylbutanoic acid, phenylbutyric acid, 4-phenylbutyrate, butyric acid, 4-phenyl, 4-phenyl-n-butyric acid
PubChem CID 4775
ChEBI CHEBI:41500
IUPAC Name 4-phenylbutanoic acid
SMILES C1=CC=C(C=C1)CCCC(=O)O

Specifications

Melting Point 49°C to 53°C
Boiling Point 165°C (10 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 638180
Solubility Information Soluble in water and methanol.
Formula Weight 164.2
Percent Purity 99%
Chemical Name or Material 4-Phenylbutyric acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 217-341-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only