missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylbutylamine Hydrochloride 98.0+%, TCI America™

Catalog No. A159025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A159025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A159025G Supplier TCI America Supplier No. A159025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 30684-06-1
Molecular Formula C10H16ClN
Molecular Weight (g/mol) 185.695
MDL Number MFCD03093616
InChI Key DGDORWAJHREUEO-UHFFFAOYSA-N
Synonym 1-Amino-4-phenylbutane Hydrochloride
PubChem CID 44630065
IUPAC Name 4-phenylbutan-1-amine;hydrochloride
SMILES C1=CC=C(C=C1)CCCCN.Cl

Specifications

Melting Point 165°C
Color White
Quantity 25 g
Formula Weight 185.70
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylbutylamine Hydrochloride