missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenylbutyl Chloride 98.0+%, TCI America™
Supplier: TCI America P138925G
Specifications
| 4-Phenylbutyl Chloride | |
| Yellow | |
| MFCD00037117 | |
| 4-chlorobutyl benzene, 1-chloro-4-phenylbutane, 4-phenylbutyl chloride, 1-4-chlorobutyl benzene, benzene, 4-chlorobutyl, phenylbutylchloride, 1-chloro4-phenylbutane, acmc-209kcr, 4-phenyl-1-chlorobutane, 4-phenyl-n-butyl chloride | |
| C1=CC=C(C=C1)CCCCCl | |
| 168.664 | |
| 168.66 | |
| Liquid |
| 4830-93-7 | |
| C10H13Cl | |
| 25 g | |
| FLLZCZIHURYEQP-UHFFFAOYSA-N | |
| 4-chlorobutylbenzene | |
| 78543 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 4830-93-7 | |
| 168.664 | |
| FLLZCZIHURYEQP-UHFFFAOYSA-N | |
| 78543 | |
| C1=CC=C(C=C1)CCCCCl |
| C10H13Cl | |
| MFCD00037117 | |
| 4-chlorobutyl benzene, 1-chloro-4-phenylbutane, 4-phenylbutyl chloride, 1-4-chlorobutyl benzene, benzene, 4-chlorobutyl, phenylbutylchloride, 1-chloro4-phenylbutane, acmc-209kcr, 4-phenyl-1-chlorobutane, 4-phenyl-n-butyl chloride | |
| 4-chlorobutylbenzene |
Safety and Handling
TSCA : No