missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylbutyl Chloride 98.0+%, TCI America™

Catalog No. p-7135112
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13895G 5 g
P138925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13895G Supplier TCI America Supplier No. P13895G
Only null left

Chemical Identifiers

CAS 4830-93-7
Molecular Formula C10H13Cl
Molecular Weight (g/mol) 168.664
MDL Number MFCD00037117
InChI Key FLLZCZIHURYEQP-UHFFFAOYSA-N
Synonym 4-chlorobutyl benzene, 1-chloro-4-phenylbutane, 4-phenylbutyl chloride, 1-4-chlorobutyl benzene, benzene, 4-chlorobutyl, phenylbutylchloride, 1-chloro4-phenylbutane, acmc-209kcr, 4-phenyl-1-chlorobutane, 4-phenyl-n-butyl chloride
PubChem CID 78543
IUPAC Name 4-chlorobutylbenzene
SMILES C1=CC=C(C=C1)CCCCCl

Specifications

Color Yellow
Quantity 5 g
Formula Weight 168.66
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Phenylbutyl Chloride