missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenylbenzoyl Chloride 98.0+%, TCI America™
Supplier: TCI America P107925G
Specifications
| 4-Phenylbenzoyl Chloride | |
| 113°C | |
| 160°C | |
| MFCD00000692 | |
| 3261 | |
| JPVUWCPKMYXOKW-UHFFFAOYSA-N | |
| [1,1'-biphenyl]-4-carbonyl chloride | |
| 84151 | |
| ≥98.0% (GC,T) |
| 14002-51-8 | |
| White | |
| C13H9ClO | |
| 25 g | |
| 4-biphenylcarbonyl chloride, 1,1'-biphenyl-4-carbonyl chloride, biphenyl-4-carbonyl chloride, p-phenylbenzoyl chloride, 4-biphenylcarbonylchloride, 4-phenyl benzoyl chloride, 4-biphenyl carbonyl chloride, 1,1'-biphenyl-4carbonyl chloride | |
| ClC(=O)C1=CC=C(C=C1)C1=CC=CC=C1 | |
| 216.66 | |
| 216.66 | |
| Crystalline Powder |
Chemical Identifiers
| 14002-51-8 | |
| 216.66 | |
| JPVUWCPKMYXOKW-UHFFFAOYSA-N | |
| 84151 | |
| ClC(=O)C1=CC=C(C=C1)C1=CC=CC=C1 |
| C13H9ClO | |
| MFCD00000692 | |
| 4-biphenylcarbonyl chloride, 1,1'-biphenyl-4-carbonyl chloride, biphenyl-4-carbonyl chloride, p-phenylbenzoyl chloride, 4-biphenylcarbonylchloride, 4-phenyl benzoyl chloride, 4-biphenyl carbonyl chloride, 1,1'-biphenyl-4carbonyl chloride | |
| [1,1'-biphenyl]-4-carbonyl chloride |
Safety and Handling
TSCA : Yes