missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylbenzoyl Chloride 98.0+%, TCI America™

Catalog No. p-7134931
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10795G 5 g
P107925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10795G Supplier TCI America Supplier No. P10795G
Only null left

Chemical Identifiers

CAS 14002-51-8
Molecular Formula C13H9ClO
Molecular Weight (g/mol) 216.66
MDL Number MFCD00000692
InChI Key JPVUWCPKMYXOKW-UHFFFAOYSA-N
Synonym 4-biphenylcarbonyl chloride, 1,1'-biphenyl-4-carbonyl chloride, biphenyl-4-carbonyl chloride, p-phenylbenzoyl chloride, 4-biphenylcarbonylchloride, 4-phenyl benzoyl chloride, 4-biphenyl carbonyl chloride, 1,1'-biphenyl-4carbonyl chloride
PubChem CID 84151
IUPAC Name [1,1'-biphenyl]-4-carbonyl chloride
SMILES ClC(=O)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 113°C
Color White
Boiling Point 160°C
Quantity 5 g
UN Number 3261
Formula Weight 216.66
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylbenzoyl Chloride