missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylazophenol, 98%

Catalog No. AAB2519822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2519822 100 g
AAB2519806 5 g
AAB2519814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2519822 Supplier Thermo Scientific Chemicals Supplier No. B2519822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1689-82-3 | C12H10N2O | 198.23 g/mol

4-Phenylazophenol is used to varnish, grease, paraffin wax, resin, soap kind of coloring. It is used in printing/solvent based writing inks. It is used for providing transparent metallic effects like gold imitation provided on metalized polyester films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenylazophenol is used to varnish, grease, paraffin wax, resin, soap kind of coloring. It is used in printing/solvent based writing inks. It is used for providing transparent metallic effects like gold imitation provided on metalized polyester films.

Solubility
Soluble in acetone, ethanol, benzene and ether. Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 1689-82-3
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00002330
InChI Key JTSBGMZPPPULTA-UHFFFAOYSA-N
Synonym 4-hydroxyazobenzene, 4-phenylazophenol, 4-phenyldiazenyl phenol, solvent yellow 7, p-phenylazophenol, 4-phenylazo phenol, p-hydroxyazobenzene, organol yellow ap, c.i. solvent yellow 7, phenol, 4-phenylazo
PubChem CID 5354198
IUPAC Name 4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one
SMILES O=C1C=CC(C=C1)=NNC1=CC=CC=C1

Specifications

Melting Point 152°C to 155°C
Quantity 100 g
Solubility Information Soluble in acetone,ethanol,benzene and ether. Insoluble in water.
Formula Weight 198.23
Percent Purity 98%
Chemical Name or Material 4-Phenylazophenol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 216-880-6
RTECSNumber SM8300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only