missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylazodiphenylamine, 96%

Catalog No. AAA1861206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1861206 5 g
AAA1861214 25 g
AAA1861222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1861206 Supplier Thermo Scientific Chemicals Supplier No. A1861206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 101-75-7 | C18H15N3 | 273.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 101-75-7
Molecular Formula C18H15N3
Molecular Weight (g/mol) 273.34
MDL Number MFCD00003023
InChI Key VXLFYNFOITWQPM-UHFFFAOYSA-N
Synonym 4-anilinoazobenzene, 4-phenylazo diphenylamine, 4-phenylazodiphenylamine, 4-benzeneazodiphenylamine, benzenamine, n-phenyl-4-phenylazo, azobenzene, 4-anilino, n-phenyl-4-aminoazobenzene, 4-phenylamino azobenzene, diphenylamine, 4-phenylazo, n-phenyl-4-phenylazo aniline
PubChem CID 7575
IUPAC Name N-phenyl-4-phenyldiazenylaniline
SMILES N(C1=CC=CC=C1)C1=CC=C(C=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 80°C to 86°C
Quantity 5 g
Formula Weight 273.34
Percent Purity 97%
Chemical Name or Material 4-Phenylazodiphenylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-972-3
RTECSNumber JK0185000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only