missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylazobenzoyl Chloride 98.0+%, TCI America™

Catalog No. p-7134447
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P01441G 1 g
P01445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P01441G Supplier TCI America Supplier No. P01441G

Chemical Identifiers

CAS 104-24-5
Molecular Formula C13H9ClN2O
Molecular Weight (g/mol) 244.68
MDL Number MFCD00041722
InChI Key RYMHZBAYPLCCAC-UHFFFAOYSA-N
PubChem CID 66910
IUPAC Name 4-(2-phenyldiazen-1-yl)benzoyl chloride
SMILES ClC(=O)C1=CC=C(C=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 95°C
Quantity 1 g
UN Number 3261
Synonym 4-phenylazobenzoyl chloride, p-phenylazobenzoyl chloride, benzoyl chloride, 4-phenylazo, p-phenylazo benzoyl chloride, 4-phenylazo benzoyl chloride, benzoyl chloride,4-2-phenyldiazenyl, 4-2-phenyldiazen-1-yl benzoyl chloride, benzoyl chloride, 4-2-phenyldiazenyl, azoyl chloride, p-phenylazobenzoylchlorid
Formula Weight 244.68
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylazobenzoyl Chloride