accessibility menu, dialog, popup

UserName

CAS RN 104-24-5

IUPAC Name: 4-(2-phenyldiazen-1-yl)benzoyl chloride
Synonyms: 4-(2-phenyldiazen-1-yl)benzoyl chloride
Molecular Weight (g/mol): 244.68
Molecular Formula: C13H9ClN2O
InChi Key: RYMHZBAYPLCCAC-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(C=C1)N=NC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results