missing translation for 'onlineSavingsMsg'
Learn More

4-(Phenylazo)benzoic Acid 98.0+%, TCI America™

Catalog No. P01431G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P01431G 1 g
P01435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P01431G Supplier TCI America Supplier No. P01431G

Chemical Identifiers

CAS 1562-93-2
Molecular Formula C13H10N2O2
Molecular Weight (g/mol) 226.235
MDL Number MFCD00020348
InChI Key CSPTZWQFHBVOLO-UHFFFAOYSA-N
Synonym Azobenzene-4-carboxylic Acid
PubChem CID 15276
IUPAC Name 4-phenyldiazenylbenzoic acid
SMILES C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 248°C
Quantity 1 g
Formula Weight 226.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Phenylazo)benzoic Acid