accessibility menu, dialog, popup

UserName

CAS RN 1562-93-2

IUPAC Name: 4-(2-phenyldiazen-1-yl)benzoic acid
Synonyms: 4-(2-phenyldiazen-1-yl)benzoic acid
Molecular Weight (g/mol): 226.24
Molecular Formula: C13H10N2O2
InChi Key: CSPTZWQFHBVOLO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)N=NC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results