missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-cyanocinnamamide matrix substance for MALDI-MS, 98.5 (HPLC), ≥98.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012654
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012654 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012654 Supplier Merck Emd Millipore Supplier No. 69028100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 116374-04-0
Linear Formula C16H12N2 O
Synonym 4-Phenyl-alpha-cyanocinnamic acid amide
Type Organic Standard
Quantity 100 mg
Formula Weight 248.28
Percent Purity ≥98.5% (HPLC)
Grade Matrix Substance for MALDI-MS