missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-1-cyclohexene 96.0+%, TCI America™

Catalog No. P14455G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14455G 5 g
P14451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14455G Supplier TCI America Supplier No. P14455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4994-16-5
Molecular Formula C12H14
Molecular Weight (g/mol) 158.244
MDL Number MFCD00049188
InChI Key XWCWNUSFQVJNDI-UHFFFAOYSA-N
Synonym 3-Cyclohexen-1-ylbenzene
PubChem CID 21096
IUPAC Name cyclohex-3-en-1-ylbenzene
SMILES C1CC(CC=C1)C2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 235°C
Quantity 5 g
Formula Weight 158.24
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Phenyl-1-cyclohexene
RTECSNumber CZ1301000
TSCA No