CAS RN 4994-16-5
IUPAC Name:
(cyclohex-3-en-1-yl)benzene
Synonyms:
4-phenylcyclohexene
Molecular Weight (g/mol):
158.24
Molecular Formula:
C12H14
InChi Key:
XWCWNUSFQVJNDI-UHFFFAOYNA-N
SMILES:
C1CC(CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
4-Phenyl-1-cyclohexene 96.0+%, TCI America™
CAS: 4994-16-5 Molecular Formula: C12H14 Molecular Weight (g/mol): 158.244 MDL Number: MFCD00049188 InChI Key: XWCWNUSFQVJNDI-UHFFFAOYSA-N Synonym: 3-Cyclohexen-1-ylbenzene PubChem CID: 21096 IUPAC Name: cyclohex-3-en-1-ylbenzene SMILES: C1CC(CC=C1)C2=CC=CC=C2