Learn More
4-Phenyl-1-butyne, 98%
CAS: 16520-62-0 | C10H10 | 130.19 g/mol
$190.66 - $621.38
Chemical Identifiers
| CAS | 16520-62-0 |
|---|---|
| Molecular Formula | C10H10 |
| Molecular Weight (g/mol) | 130.19 |
| MDL Number | MFCD00041667 |
| InChI Key | QDEOKXOYHYUKMS-UHFFFAOYSA-N |
| Synonym | 4-phenyl-1-butyne, 3-butynylbenzene, but-3-yn-1-ylbenzene, benzene, 3-butynyl, 2-phenylethylacetylene, ccris 7100, phenethylacetylene, 4-phenyl-1-butyn, 3-butynylbenzene #, 4-phenylbut-1-yne |
| PubChem CID | 123360 |
| IUPAC Name | but-3-ynylbenzene |
| SMILES | C#CCCC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1294606
|
Thermo Scientific Chemicals
L1294606 |
5 g |
Each for $190.66
|
|
|||||
|
AAL1294614
|
Thermo Scientific Chemicals
L1294614 |
25 g |
Each for $621.38
|
|
|||||
Description
4-Phenyl-1-butyne is used as an intermediate in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Phenyl-1-butyne is used as an intermediate in chemical research.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents.
Chemical Identifiers
| 16520-62-0 | |
| 130.19 | |
| QDEOKXOYHYUKMS-UHFFFAOYSA-N | |
| 123360 | |
| C#CCCC1=CC=CC=C1 |
| C10H10 | |
| MFCD00041667 | |
| 4-phenyl-1-butyne, 3-butynylbenzene, but-3-yn-1-ylbenzene, benzene, 3-butynyl, 2-phenylethylacetylene, ccris 7100, phenethylacetylene, 4-phenyl-1-butyn, 3-butynylbenzene #, 4-phenylbut-1-yne | |
| but-3-ynylbenzene |
Specifications
| 16520-62-0 | |
| 189°C to 190°C | |
| C6H5(CH2)2C≡CH | |
| MFCD00041667 | |
| 1362221 | |
| Not miscible or difficult to mix in water. | |
| C#CCCC1=CC=CC=C1 | |
| 130.19 | |
| 130.19 | |
| 4-Phenyl-1-butyne |
| 0.926 | |
| C10H10 | |
| 1.521 | |
| 5 g | |
| 4-phenyl-1-butyne, 3-butynylbenzene, but-3-yn-1-ylbenzene, benzene, 3-butynyl, 2-phenylethylacetylene, ccris 7100, phenethylacetylene, 4-phenyl-1-butyn, 3-butynylbenzene #, 4-phenylbut-1-yne | |
| QDEOKXOYHYUKMS-UHFFFAOYSA-N | |
| but-3-ynylbenzene | |
| 123360 | |
| 98% |
Safety and Handling
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only