missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$106.25 - $106.25
Specifications
| CAS | 51067-38-0 |
|---|---|
| Melting Point | 115°C |
| Molecular Formula | C12H11BO3 |
| MDL Number | MFCD00093312 |
| InChI Key | KFXUHRXGLWUOJT-UHFFFAOYSA-N |
Specifications
| 51067-38-0 | |
| C12H11BO3 | |
| KFXUHRXGLWUOJT-UHFFFAOYSA-N | |
| 2734377 | |
| Crystalline Powder |
| 115°C | |
| MFCD00093312 | |
| (4-phenoxyphenyl)boronic acid | |
| 214.03 |