missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. P19745G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P19745G Supplier TCI America Supplier No. P19745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-Phenoxyphenylboronic Acid (contains varying amounts of Anhydride)
CAS 51067-38-0
Melting Point 115°C
Molecular Formula C12H11BO3
Quantity 5 g
MDL Number MFCD00093312
Synonym 4-phenoxyphenyl boronic acid, 4-phenoxybenzeneboronic acid, 4-phenoxyphenyl boranediol, boronic acid, 4-phenoxyphenyl, p-phenoxyphenyl boronic acid, pubchem7890, acmc-209krn, phenoxyphenylboronic acid, d0d2ja
InChI Key KFXUHRXGLWUOJT-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(OC2=CC=CC=C2)C=C1
IUPAC Name (4-phenoxyphenyl)boronic acid
Molecular Weight (g/mol) 214.03
PubChem CID 2734377
Formula Weight 214.03
Physical Form Crystalline Powder
Show More Show Less