missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenoxyphenol 99.0+%, TCI America™
$121.28 - $1180.13
Chemical Identifiers
| CAS | 831-82-3 |
|---|---|
| Molecular Formula | C12H10O2 |
| Molecular Weight (g/mol) | 186.21 |
| MDL Number | MFCD00002331 |
| InChI Key | ZSBDGXGICLIJGD-UHFFFAOYSA-N |
| Synonym | p-phenoxyphenol, 4-hydroxydiphenyl ether, phenol, 4-phenoxy, p-hydroxydiphenyl ether, phenol, p-phenoxy, hydroquinone monophenyl ether, unii-d83r742gjr, 4-phenyloxy phenol, 4-phenoxy phenol, 4-phenoxy-phenol |
| PubChem CID | 13254 |
| ChEBI | CHEBI:39264 |
| IUPAC Name | 4-phenoxyphenol |
| SMILES | C1=CC=C(C=C1)OC2=CC=C(C=C2)O |
Chemical Identifiers
| 831-82-3 | |
| 186.21 | |
| ZSBDGXGICLIJGD-UHFFFAOYSA-N | |
| 13254 | |
| 4-phenoxyphenol |
| C12H10O2 | |
| MFCD00002331 | |
| p-phenoxyphenol, 4-hydroxydiphenyl ether, phenol, 4-phenoxy, p-hydroxydiphenyl ether, phenol, p-phenoxy, hydroquinone monophenyl ether, unii-d83r742gjr, 4-phenyloxy phenol, 4-phenoxy phenol, 4-phenoxy-phenol | |
| CHEBI:39264 | |
| C1=CC=C(C=C1)OC2=CC=C(C=C2)O |
Specifications
| 831-82-3 | |
| 167°C | |
| MFCD00002331 | |
| p-phenoxyphenol, 4-hydroxydiphenyl ether, phenol, 4-phenoxy, p-hydroxydiphenyl ether, phenol, p-phenoxy, hydroquinone monophenyl ether, unii-d83r742gjr, 4-phenyloxy phenol, 4-phenoxy phenol, 4-phenoxy-phenol | |
| C1=CC=C(C=C1)OC2=CC=C(C=C2)O | |
| 186.21 | |
| CHEBI:39264 | |
| ≥99.0% (GC) | |
| 4-Phenoxyphenol |
| 83°C | |
| C12H10O2 | |
| 25 g | |
| ZSBDGXGICLIJGD-UHFFFAOYSA-N | |
| 4-phenoxyphenol | |
| 13254 | |
| 186.21 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-4070
TSCA : Yes