missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxyphenol 99.0+%, TCI America™

Catalog No. P0950500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0950500G 500 g
P095025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0950500G Supplier TCI America Supplier No. P0950500G
Only null left

Chemical Identifiers

CAS 831-82-3
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00002331
InChI Key ZSBDGXGICLIJGD-UHFFFAOYSA-N
Synonym p-phenoxyphenol, 4-hydroxydiphenyl ether, phenol, 4-phenoxy, p-hydroxydiphenyl ether, phenol, p-phenoxy, hydroquinone monophenyl ether, unii-d83r742gjr, 4-phenyloxy phenol, 4-phenoxy phenol, 4-phenoxy-phenol
PubChem CID 13254
ChEBI CHEBI:39264
IUPAC Name 4-phenoxyphenol
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)O

Specifications

Melting Point 83°C
Boiling Point 167°C
Quantity 500 g
Formula Weight 186.21
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenoxyphenol
EINECSNumber (3)-4070
TSCA Yes