missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxybenzyl bromide, 97%

Catalog No. p-7032438
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33895MD 250 mg
AAH3389503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33895MD Supplier Thermo Scientific Chemicals Supplier No. H33895MD
Only null left

CAS: 36881-42-2 | C13H11BrO | 263.134 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36881-42-2
Molecular Formula C13H11BrO
Molecular Weight (g/mol) 263.134
MDL Number MFCD04038693
InChI Key CPIGBCFBFZSCQI-UHFFFAOYSA-N
PubChem CID 1514251
IUPAC Name 1-(bromomethyl)-4-phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)CBr

Specifications

Quantity 250 mg
UN Number UN3265
Synonym 1-bromomethyl-4-phenoxybenzene, 4-phenoxybenzyl bromide, 1-bromomethyl-4-phenoxy-benzene, benzene,1-bromomethyl-4-phenoxy, 4-phenyloxybenzylbromide, 4-bromomethyl-1-phenoxybenzene, benzene, 1-bromomethyl-4-phenoxy
Formula Weight 263.13
Percent Purity 97%
Chemical Name or Material 4-Phenoxybenzyl bromide
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 253-253-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only