missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxybenzyl alcohol, 97%

Catalog No. AAH3313803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3313803 1 g
AAH3313806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3313803 Supplier Thermo Scientific Chemicals Supplier No. H3313803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2215-78-3 | C13H12O2 | 200.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2215-78-3
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.24
MDL Number MFCD01463970
InChI Key FEOMFFKZOZMBKD-UHFFFAOYSA-N
PubChem CID 826195
IUPAC Name (4-phenoxyphenyl)methanol
SMILES OCC1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Quantity 1 g
Synonym 4-phenoxyphenyl methanol, 4-phenoxybenzyl alcohol, 4-phenoxy-phenyl-methanol, 4-phenoxybenzylalcohol, p-phenoxybenzyl alcohol, benzenemethanol,4-phenoxy, benzenemethanol, 4-phenoxy, 4-phenyloxy phenyl methanol, 4-phenoxyphenyl methan-1-ol
Formula Weight 200.24
Percent Purity 97%
Chemical Name or Material 4-Phenoxybenzyl alcohol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only