missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxybenzonitrile, 96%

CAS: 3096-81-9 | C13H9NO | 195.221 g/mol

$52.44 - $175.12

Chemical Identifiers

CAS 3096-81-9
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.221
MDL Number MFCD00017346
InChI Key UYHCIOZMFCLUDP-UHFFFAOYSA-N
Synonym benzonitrile, 4-phenoxy, 4-phenoxybenzenecarbonitrile, 4-cyanodiphenyl ether, acmc-209hjp, 4-phenoxybenzonitrile, ksc497e6d
PubChem CID 137821
IUPAC Name 4-phenoxybenzonitrile
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2536404
View Documents
Thermo Scientific Chemicals
B2536404
2 g
Each for $52.44
Only null left
 
AAB2536409
View Documents
Thermo Scientific Chemicals
B2536409
10 g
Each for $175.12
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3096-81-9
195.221
UYHCIOZMFCLUDP-UHFFFAOYSA-N
137821
C1=CC=C(C=C1)OC2=CC=C(C=C2)C#N
C13H9NO
MFCD00017346
benzonitrile, 4-phenoxy, 4-phenoxybenzenecarbonitrile, 4-cyanodiphenyl ether, acmc-209hjp, 4-phenoxybenzonitrile, ksc497e6d
4-phenoxybenzonitrile

Specifications

3096-81-9
>110°C (230°F)
MFCD00017346
UN3439
UYHCIOZMFCLUDP-UHFFFAOYSA-N
4-phenoxybenzonitrile
137821
96%
46°C to 48°C
C13H9NO
2 g
benzonitrile, 4-phenoxy, 4-phenoxybenzenecarbonitrile, 4-cyanodiphenyl ether, acmc-209hjp, 4-phenoxybenzonitrile, ksc497e6d
C1=CC=C(C=C1)OC2=CC=C(C=C2)C#N
195.221
195.22
4-Phenoxybenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only