missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxybenzonitrile, 96%

Catalog No. AAB2536409
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2536409 10 g
AAB2536404 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2536409 Supplier Thermo Scientific Chemicals Supplier No. B2536409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3096-81-9 | C13H9NO | 195.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3096-81-9
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.221
MDL Number MFCD00017346
InChI Key UYHCIOZMFCLUDP-UHFFFAOYSA-N
Synonym benzonitrile, 4-phenoxy, 4-phenoxybenzenecarbonitrile, 4-cyanodiphenyl ether, acmc-209hjp, 4-phenoxybenzonitrile, ksc497e6d
PubChem CID 137821
IUPAC Name 4-phenoxybenzonitrile
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)C#N

Specifications

Melting Point 46°C to 48°C
Flash Point >110°C (230°F)
Quantity 10 g
UN Number UN3439
Formula Weight 195.22
Percent Purity 96%
Chemical Name or Material 4-Phenoxybenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only