missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxyaniline, 97%

Catalog No. AAA1604814
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1604814 25 g
AAA1604822 100 g
AAA1604836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1604814 Supplier Thermo Scientific Chemicals Supplier No. A1604814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 139-59-3 | C12H11NO | 185.226 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 139-59-3
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.226
MDL Number MFCD00007862
InChI Key WOYZXEVUWXQVNV-UHFFFAOYSA-N
Synonym p-phenoxyaniline, 4-aminodiphenyl ether, benzenamine, 4-phenoxy, 4-phenoxybenzenamine, 4-aminophenyl phenyl ether, 4-aminodifenylether, 4-amino-1-phenoxybenzene, aniline, p-phenoxy, 4-aminobiphenyl ether, 4-aminodiphenylether
PubChem CID 8764
IUPAC Name 4-phenoxyaniline
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)N

Specifications

Melting Point 82°C to 86°C
Boiling Point 140°C (2 mmHg)
Quantity 25 g
UN Number UN3077
Beilstein 777708
Formula Weight 185.23
Percent Purity 97%
Chemical Name or Material 4-Phenoxyaniline
Hazard Category H302-H315-H317-H319-H335
Hazard Statement GHS H Statement
H301-H341-H319-H317
Toxic if swallowed.
Suspected of causing genetic defects.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 205-367-2
RTECSNumber BY7930000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only