missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxyaniline, 97%

Catalog No. AC130235000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC130235000 500 g Glass bottle
AC130230050 5 g Glass bottle
AC130231000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC130235000 Supplier Thermo Scientific Chemicals Supplier No. 130235000
Only null left

CAS: 139-59-3 | C12H11NO | 185.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 139-59-3
MDL Number MFCD00007862
InChI Key WOYZXEVUWXQVNV-UHFFFAOYSA-N
Synonym p-phenoxyaniline, 4-aminodiphenyl ether, benzenamine, 4-phenoxy, 4-phenoxybenzenamine, 4-aminophenyl phenyl ether, 4-aminodifenylether, 4-amino-1-phenoxybenzene, aniline, p-phenoxy, 4-aminobiphenyl ether, 4-aminodiphenylether
PubChem CID 8764
IUPAC Name 4-phenoxyaniline
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 82°C to 86°C
Color Brown to Brown-Green
Boiling Point 140°C (3.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C12H11NO
Linear Formula C6H5OC6H4NH2
Quantity 500 g
Beilstein 13, 438
Solubility Information Solubility in water: < 1g/l in water (20°C). Other solubilities: soluble in isopropanol,toluene,very soluble in acetone,1,2-dichloroethane
Molecular Weight (g/mol) 185.23
Formula Weight 185.23
Percent Purity 97%
Physical Form Crystalline Flakes or Powder
Chemical Name or Material 4-Phenoxyaniline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF IN EYES: Rinse
EINECSNumber 205-367-2

RUO – Research Use Only