missing translation for 'onlineSavingsMsg'
Learn More

4'-Phenoxyacetophenone, 98+%

Catalog No. AAA1239222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1239222 100 g
AAA1239214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1239222 Supplier Thermo Scientific Chemicals Supplier No. A1239222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5031-78-7 | C14H12O2 | 212.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5031-78-7
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00008744
InChI Key DJNIFZYQFLFGDT-UHFFFAOYSA-N
Synonym 4'-phenoxyacetophenone, 1-4-phenoxyphenyl ethanone, 4-phenoxyacetophenone, p-phenoxyacetophenone, 1-4-phenoxyphenyl ethan-1-one, ethanone, 1-4-phenoxyphenyl, 4-acetyldiphenyl ether, 1-4-phenoxyphenyl-1-ethanone, 1-4-phenoxyphenyl ethanon, pubchem13195
PubChem CID 236783
SMILES CC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Melting Point 48°C to 52°C
Boiling Point 325°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 2049295
IUPAC Name 1-(4-phenoxyphenyl)ethan-1-one
Formula Weight 212.25
Percent Purity ≥98%
Chemical Name or Material 4'-Phenoxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only