missing translation for 'onlineSavingsMsg'
Learn More

4-Pentynoic Acid 98.0+%, TCI America™

Catalog No. P23411G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23411G 1 g
P23415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23411G Supplier TCI America Supplier No. P23411G

Chemical Identifiers

CAS 6089-09-4
Molecular Formula C5H5O2
Molecular Weight (g/mol) 97.09
MDL Number MFCD00004407
InChI Key MLBYLEUJXUBIJJ-UHFFFAOYSA-M
Synonym 4-pentynoic acid, propargylacetic acid, pent-4-yn-1-oic acid, 4-pentynoicacid, pent4-ynoic acid, zlchem 796, pubchem3491, pent-4-yn-oic acid, acmc-1az7d
PubChem CID 22464
IUPAC Name pent-4-ynoic acid
SMILES C#CCCC(=O)O

Specifications

Melting Point 57°C
Color White-Yellow
Boiling Point 204°C
Quantity 1 g
UN Number 3261
Formula Weight 98.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Pentynoic Acid
RTECSNumber SC4751000
TSCA No
Recommended Storage Refrigerator