accessibility menu, dialog, popup

UserName

OOCH

CAS RN 6089-09-4

IUPAC Name: pent-4-ynoate
Synonyms: pent-4-ynoate
Molecular Weight (g/mol): 97.09
Molecular Formula: C5H5O2
InChi Key: MLBYLEUJXUBIJJ-UHFFFAOYSA-M
SMILES: [O-]C(=O)CCC#C
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)