missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pentenyl Acetate 98.0+%, TCI America™
$139.81 - $1092.33
Chemical Identifiers
| CAS | 1576-85-8 |
|---|---|
| Molecular Formula | C7H12O2 |
| Molecular Weight (g/mol) | 128.17 |
| MDL Number | MFCD00008723 |
| InChI Key | LVHDNIMNOMRZMF-UHFFFAOYSA-N |
| Synonym | 4-pentenyl acetate, 4-penten-1-yl acetate, 5-acetoxy-1-pentene, unii-v4sif0mabv, pent-4-en-1-yl acetate, pent-4-ene-1-yl acetate, ch3c o o ch2 3ch=ch2, v4sif0mabv, 4-penten-1-ol acetate, 4-pentenyl acetate # |
| PubChem CID | 74096 |
| ChEBI | CHEBI:87515 |
| IUPAC Name | pent-4-en-1-yl acetate |
| SMILES | CC(=O)OCCCC=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A134525ML
|
TCI America
A134525ML |
25 mL |
Each for $139.81
|
|
|||||
|
A1345500ML
|
TCI America
A1345500ML |
500 mL |
Each for $1,092.33
|
|
|||||
Chemical Identifiers
| 1576-85-8 | |
| 128.17 | |
| LVHDNIMNOMRZMF-UHFFFAOYSA-N | |
| 74096 | |
| pent-4-en-1-yl acetate |
| C7H12O2 | |
| MFCD00008723 | |
| 4-pentenyl acetate, 4-penten-1-yl acetate, 5-acetoxy-1-pentene, unii-v4sif0mabv, pent-4-en-1-yl acetate, pent-4-ene-1-yl acetate, ch3c o o ch2 3ch=ch2, v4sif0mabv, 4-penten-1-ol acetate, 4-pentenyl acetate # | |
| CHEBI:87515 | |
| CC(=O)OCCCC=C |
Specifications
| 1576-85-8 | |
| 151°C | |
| MFCD00008723 | |
| 3272 | |
| LVHDNIMNOMRZMF-UHFFFAOYSA-N | |
| pent-4-en-1-yl acetate | |
| 74096 | |
| 128.17 | |
| Liquid |
| Colorless | |
| C7H12O2 | |
| 25 mL | |
| 4-pentenyl acetate, 4-penten-1-yl acetate, 5-acetoxy-1-pentene, unii-v4sif0mabv, pent-4-en-1-yl acetate, pent-4-ene-1-yl acetate, ch3c o o ch2 3ch=ch2, v4sif0mabv, 4-penten-1-ol acetate, 4-pentenyl acetate # | |
| CC(=O)OCCCC=C | |
| 128.17 | |
| CHEBI:87515 | |
| ≥98.0% (GC) | |
| 4-Pentenyl Acetate |
Safety and Handling
EINECSNumber : (2)-0760
TSCA : No