accessibility menu, dialog, popup

UserName

H3COOCH2

CAS RN 1576-85-8

IUPAC Name: pent-4-en-1-yl acetate
Synonyms: 4-penten-1-yl acetate
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: LVHDNIMNOMRZMF-UHFFFAOYSA-N
SMILES: CC(=O)OCCCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results