missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pentenyl Acetate 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 151°C |
| Quantity | 500 mL |
| UN Number | 3272 |
| Formula Weight | 128.17 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 4-Pentenyl Acetate |
Safety and Handling
| EINECSNumber | (2)-0760 |
| TSCA | No |