missing translation for 'onlineSavingsMsg'
Learn More

4-Pentenyl Acetate 98.0+%, TCI America™

Catalog No. A134525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A134525ML 25 mL
A1345500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A134525ML Supplier TCI America Supplier No. A134525ML
Only null left

Chemical Identifiers

CAS 1576-85-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00008723
InChI Key LVHDNIMNOMRZMF-UHFFFAOYSA-N
Synonym 4-pentenyl acetate, 4-penten-1-yl acetate, 5-acetoxy-1-pentene, unii-v4sif0mabv, pent-4-en-1-yl acetate, pent-4-ene-1-yl acetate, ch3c o o ch2 3ch=ch2, v4sif0mabv, 4-penten-1-ol acetate, 4-pentenyl acetate #
PubChem CID 74096
ChEBI CHEBI:87515
IUPAC Name pent-4-en-1-yl acetate
SMILES CC(=O)OCCCC=C

Specifications

Color Colorless
Boiling Point 151°C
Quantity 25 mL
UN Number 3272
Formula Weight 128.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Pentenyl Acetate
EINECSNumber (2)-0760
TSCA No