missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pentenoic Acid 98.0+%, TCI America™
$152.08 - $454.34
Chemical Identifiers
| CAS | 591-80-0 |
|---|---|
| Molecular Formula | C5H8O2 |
| Molecular Weight (g/mol) | 100.12 |
| MDL Number | MFCD00004408 |
| InChI Key | HVAMZGADVCBITI-UHFFFAOYSA-N |
| Synonym | 4-pentenoic acid, allylacetic acid, allyl acetic acid, 4 pa, 3-vinylpropionic acid, delta 4-pentenoic acid, unii-d4s77y29fb, fema no. 2843, .delta.4-pentenoic acid, wln: qv3u1 |
| PubChem CID | 61138 |
| ChEBI | CHEBI:35936 |
| IUPAC Name | pent-4-enoic acid |
| SMILES | OC(=O)CCC=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P064525ML
|
TCI America
P064525ML |
25 mL |
Each for $152.08
|
|
|||||
|
P0645100ML
|
TCI America
P0645100ML |
100 mL |
Each for $454.34
|
|
|||||
Chemical Identifiers
| 591-80-0 | |
| 100.12 | |
| HVAMZGADVCBITI-UHFFFAOYSA-N | |
| 61138 | |
| pent-4-enoic acid |
| C5H8O2 | |
| MFCD00004408 | |
| 4-pentenoic acid, allylacetic acid, allyl acetic acid, 4 pa, 3-vinylpropionic acid, delta 4-pentenoic acid, unii-d4s77y29fb, fema no. 2843, .delta.4-pentenoic acid, wln: qv3u1 | |
| CHEBI:35936 | |
| OC(=O)CCC=C |
Specifications
| 591-80-0 | |
| 182°C | |
| MFCD00004408 | |
| 3265 | |
| HVAMZGADVCBITI-UHFFFAOYSA-N | |
| pent-4-enoic acid | |
| 61138 | |
| 100.12 | |
| Liquid |
| Yellow | |
| C5H8O2 | |
| 25 mL | |
| 4-pentenoic acid, allylacetic acid, allyl acetic acid, 4 pa, 3-vinylpropionic acid, delta 4-pentenoic acid, unii-d4s77y29fb, fema no. 2843, .delta.4-pentenoic acid, wln: qv3u1 | |
| OC(=O)CCC=C | |
| 100.12 | |
| CHEBI:35936 | |
| ≥98.0% (GC,T) | |
| 4-Pentenoic Acid |
Safety and Handling
EINECSNumber : (2)-0609
RTECSNumber : SB2800000
TSCA : Yes