accessibility menu, dialog, popup

UserName

CAS RN 591-80-0

IUPAC Name: pent-4-enoic acid
Synonyms: 4-pentenoic acid|allyl acetic acid|4-pentensaeure|allylessigsaeure|delta 4-pentenoic acid|delta(4)-pentenoic acid
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: HVAMZGADVCBITI-UHFFFAOYSA-N
SMILES: OC(=O)CCC=C
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)