missing translation for 'onlineSavingsMsg'
Learn More

4-Pentenoic acid, 99%

CAS: 591-80-0 | C5H8O2 | 100.12 g/mol

Supplier:  Thermo Scientific Chemicals 269020250

 View more versions of this product

Catalog No. AC269020250

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Pentenoic acid
-22.5°C
0.9750g/mL
89°C
98.5% min. (GC)
C5H8O2
H2C=CHCH2CH2CO2H
25 g
0.975
Solubility in water: insoluble
OC(=O)CCC=C
100.12
CHEBI:35936
99%
591-80-0
Colorless to Yellow
83.0°C to 84.0°C (12.0 mmHg)
Authentic
Glass bottle
1.4273 to 1.4303
MFCD00004408
02,425
4-pentenoic acid, allylacetic acid, allyl acetic acid, 4 pa, 3-vinylpropionic acid, delta 4-pentenoic acid, unii-d4s77y29fb, fema no. 2843, .delta.4-pentenoic acid, wln: qv3u1
HVAMZGADVCBITI-UHFFFAOYSA-N
pent-4-enoic acid
61138
100.2
Liquid

Chemical Identifiers

591-80-0
100.12
HVAMZGADVCBITI-UHFFFAOYSA-N
61138
pent-4-enoic acid
C5H8O2
MFCD00004408
4-pentenoic acid, allylacetic acid, allyl acetic acid, 4 pa, 3-vinylpropionic acid, delta 4-pentenoic acid, unii-d4s77y29fb, fema no. 2843, .delta.4-pentenoic acid, wln: qv3u1
CHEBI:35936
OC(=O)CCC=C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 209-732-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only