missing translation for 'onlineSavingsMsg'
Learn More

4-Penten-1-yl Isothiocyanate 95.0+%, TCI America™

Catalog No. I04445G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04445G 5 g
I04441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04445G Supplier TCI America Supplier No. I04445G
Only null left

Chemical Identifiers

CAS 18060-79-2
Molecular Formula C6H9NS
Molecular Weight (g/mol) 127.21
MDL Number MFCD00191527
InChI Key DBISBKDNOKIADM-UHFFFAOYSA-N
Synonym Isothiocyanic Acid 4-Penten-1-yl Ester
PubChem CID 87436
IUPAC Name 5-isothiocyanatopent-1-ene
SMILES C=CCCCN=C=S

Specifications

Color Yellow
Boiling Point 194°C
Quantity 5 g
UN Number 1992
Formula Weight 127.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Penten-1-yl Isothiocyanate