missing translation for 'onlineSavingsMsg'
Learn More

4-(Pentafluorothio)phenol, 97%

Catalog No. AAH3341306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3341306 5 g
AAH3341303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3341306 Supplier Thermo Scientific Chemicals Supplier No. H3341306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 774-94-7 | C6H5F5OS | 220.16 g/mol

Synthesis of hexylamides using fluorine derivatives as intermediate active esters. Various non-metallic derivatives of pentafluorothiophenol, containing sulfur bonded to boron, carbon, silicon, germanium, phosphorus, arsenic, antimony, sulfur, and chlorine, can be prepared.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Synthesis of hexylamides using fluorine derivatives as intermediate active esters. Various non-metallic derivatives of pentafluorothiophenol, containing sulfur bonded to boron, carbon, silicon, germanium, phosphorus, arsenic, antimony, sulfur, and chlorine, can be prepared.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 774-94-7
Molecular Formula C6H5F5OS
Molecular Weight (g/mol) 220.16
MDL Number MFCD03788516
InChI Key XHJLGVIUMCBMHL-UHFFFAOYSA-N
Synonym 4-pentafluorothio phenol, 4-pentafluorosulfanyl phenol, 4-hydroxyphenylsulfur pentafluoride, 4-pentafluoro-$l^ 6-sulfanyl phenol, 4-pentafluoro-lambda∼6∼-sulfanyl phenol, sulfur,pentafluoro 4-hydroxyphenyl-, oc-6-21-9ci, 4-pentafluorosulfanylphenol, 4-pentafluorosulfur phenol, 4-pentafluorosulphanyl phenol, 4-pentafluorothio vi phenol
PubChem CID 2779203
IUPAC Name 4-(pentafluoro-$l^{6}-sulfanyl)phenol
SMILES OC1=CC=C(C=C1)S(F)(F)(F)(F)F

Specifications

Quantity 5 g
Formula Weight 220.16
Percent Purity 97%
Chemical Name or Material 4-(Pentafluorothio)phenol
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only