missing translation for 'onlineSavingsMsg'
Learn More

4-(p-Tolyl)cyclohexanone 98.0+%, TCI America™

Catalog No. T34181G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T34181G Supplier TCI America Supplier No. T34181G
Only null left

Chemical Identifiers

CAS 40503-90-0
Molecular Formula C13H16O
Molecular Weight (g/mol) 188.27
MDL Number MFCD14584461
InChI Key XDHMORMQXOUOHT-UHFFFAOYSA-N
Synonym 4-(4-Methylphenyl)cyclohexanone
PubChem CID 14549255
IUPAC Name 4-(4-methylphenyl)cyclohexan-1-one
SMILES CC1=CC=C(C=C1)C2CCC(=O)CC2

Specifications

Melting Point 51°C
Color White
Quantity 1 g
Formula Weight 188.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(p-Tolyl)cyclohexanone