missing translation for 'onlineSavingsMsg'
Learn More

4-(p-Tolyl)butyric Acid 98.0+%, TCI America™

Catalog No. T17271G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17271G 1 g
T17275G 5 g
T172725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T17271G Supplier TCI America Supplier No. T17271G

Chemical Identifiers

CAS 4521-22-6
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00021786
InChI Key IXWOVMRDYFFXGI-UHFFFAOYSA-N
Synonym 4-p-tolyl butanoic acid, 4-p-tolyl butyric acid, 4-4-methylphenyl butanoic acid, benzenebutanoic acid, 4-methyl, 4-p-tolylbutyric acid, 4-para-tolyl-butyric acid, 4-4-methylphenyl butyric acid, 4-p-tolyl butanoicacid, 4-p-tolylbutanoic acid, 4-p-tolyl-butyric acid
PubChem CID 78279
IUPAC Name 4-(4-methylphenyl)butanoic acid
SMILES CC1=CC=C(C=C1)CCCC(=O)O

Specifications

Melting Point 58°C
Color White-Yellow
Boiling Point 150°C
Quantity 1 g
Formula Weight 178.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(p-Tolyl)butyric Acid