missing translation for 'onlineSavingsMsg'
Learn More

4-(p-Tolyl)butyric acid, 97%

Catalog No. p-7022075
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1468003 1 g
AAA1468006 5 g
AAA1468014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1468003 Supplier Thermo Scientific Chemicals Supplier No. A1468003

CAS: 4521-22-6 | C11H14O2 | 178.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4521-22-6
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00021786
InChI Key IXWOVMRDYFFXGI-UHFFFAOYSA-N
Synonym 4-p-tolyl butanoic acid, 4-p-tolyl butyric acid, 4-4-methylphenyl butanoic acid, benzenebutanoic acid, 4-methyl, 4-p-tolylbutyric acid, 4-para-tolyl-butyric acid, 4-4-methylphenyl butyric acid, 4-p-tolyl butanoicacid, 4-p-tolylbutanoic acid, 4-p-tolyl-butyric acid
PubChem CID 78279
IUPAC Name 4-(4-methylphenyl)butanoic acid
SMILES CC1=CC=C(C=C1)CCCC(=O)O

Specifications

Melting Point 55°C to 59°C
Quantity 1 g
Beilstein 639705
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material 4-(p-Tolyl)butyric acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 224-848-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only