missing translation for 'onlineSavingsMsg'
Learn More

4-Oxoretinoic acid, ≥90.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014092
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014092 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014092 Supplier Merck Emd Millipore Supplier No. 0755825MG
Only null left

Specifications

CAS 38030-57-8
Linear Formula C20H26O3
MDL Number MFCD00870472
Synonym (2 E,4 E,6 E,8 E)-3,7-Dimethyl-9-(2,6,6-trimethyl-3-oxo-1-cyclohexen-1-yl)-2,4,6,8-nonatetraenoic acid; 4-Ketoretinoic acid; 4-Oxo-all-trans-retinoic acid
Type Analytical Standard
Quantity 25 mg
Formula Weight 314.42
Percent Purity ≥90.0% (HPLC)
Grade Analytical Standard