missing translation for 'onlineSavingsMsg'
Learn More

4-Octylbenzaldehyde, 97%

Catalog No. AC368880050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC368880050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC368880050 Supplier Thermo Scientific Chemicals Supplier No. 368880050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 49763-66-8 | C15H22O | 218.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 49763-66-8
Molecular Formula C15H22O
Molecular Weight (g/mol) 218.34
MDL Number MFCD00041872
InChI Key IHKVZPVHTKOSLW-UHFFFAOYSA-N
Synonym benzaldehyde, 4-octyl, n-octylbenzaldehyde, 4-n-octylbenzaldehyde, p-octylbenzaldehyde, paraoctylbenzaldehyde, p-n-octylbenzaldehyde, para-octylbenzaldehyde, p-octylbenzaldehyde, l
PubChem CID 2734685
IUPAC Name 4-octylbenzaldehyde
SMILES CCCCCCCCC1=CC=C(C=C1)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CHOC6H4CH2(CH2)6CH3
Refractive Index 1.5100 to 1.513
Quantity 5 g
Formula Weight 218.34
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Octylbenzaldehyde, 97%
Show More Show Less

RUO – Research Use Only