missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Octanol 97.0+%, TCI America™
Supplier: TCI America O01555ML
Specifications
| 4-Octanol | |
| -42°C | |
| 176°C | |
| MFCD00014409 | |
| 4-octanol, n-octan-4-ol, 4-octanol, s, 4-octyl alcohol, butylpropylcarbinol, acmc-1az8y, 4-octanol gc | |
| CCCCC(O)CCC | |
| 130.23 | |
| 130.23 | |
| Liquid |
| 589-62-8 | |
| Colorless | |
| C8H18O | |
| 5 mL | |
| WOFPPJOZXUTRAU-UHFFFAOYNA-N | |
| octan-4-ol | |
| 11515 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 589-62-8 | |
| 130.23 | |
| WOFPPJOZXUTRAU-UHFFFAOYNA-N | |
| 11515 | |
| CCCCC(O)CCC |
| C8H18O | |
| MFCD00014409 | |
| 4-octanol, n-octan-4-ol, 4-octanol, s, 4-octyl alcohol, butylpropylcarbinol, acmc-1az8y, 4-octanol gc | |
| octan-4-ol |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : No