missing translation for 'onlineSavingsMsg'
Learn More

4-O-Benzyl-D-glucal, 97%, Thermo Scientific Chemicals

Catalog No. AAH6067803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6067803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6067803 Supplier Thermo Scientific Chemicals Supplier No. H6067803

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.

Chemical Identifiers

CAS 58871-11-7
Molecular Formula C13H16O4
Molecular Weight (g/mol) 236.27
MDL Number MFCD01863610
InChI Key OIWQZGBSQDJVJN-UPJWGTAASA-N
Synonym 4-o-benzyl-d-glucal, 2r,3s,4r-3-benzyloxy-2-hydroxymethyl-3,4-dihydro-2h-pyran-4-ol, 4-o-benzyl-d-glucal,, 1,2-dideoxy-4-o-benzyl-d-arabino-1-hexenopyranose, 2r,3s,4r-2-hydroxymethyl-3-phenylmethoxy-3,4-dihydro-2h, 2r,3s,4r-2-hydroxymethyl-3-phenylmethoxy-3,4-dihydro-2h-pyran-4-ol, 5s,4r,6r-6-hydroxymethyl-5-phenylmethoxy-4h-5,6-dihydropyran-4-ol
PubChem CID 2734727
IUPAC Name (2R,3S,4R)-2-(hydroxymethyl)-3-phenylmethoxy-3,4-dihydro-2H-pyran-4-ol
SMILES OC[C@H]1OC=C[C@@H](O)[C@@H]1OCC1=CC=CC=C1

Specifications

Melting Point 99°C to 103°C
Quantity 1 g
Optical Rotation +10° (c=1 in Chloroform)
Formula Weight 236.26
Percent Purity 97%
Chemical Name or Material 4-O-Benzyl-D-glucal
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only