missing translation for 'onlineSavingsMsg'
Learn More

4-Nonylphenol (mixture of branched chain isomers), TCI America™

Catalog No. N030025ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N030025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N030025ML Supplier TCI America Supplier No. N030025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84852-15-3
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.356
MDL Number MFCD00002396
InChI Key SNQQPOLDUKLAAF-UHFFFAOYSA-N
Synonym phenol, 2-nonyl, nonyl phenol, o-nonylphenol, nonyl, phenol,nonyl, 2-n-nonyl-phenol, ortho-n-nonyl-phenol, acmc-209t6o, bidd:er0079, nonylphenol, mixture of isomers
PubChem CID 67296
IUPAC Name 2-nonylphenol
SMILES CCCCCCCCCC1=CC=CC=C1O

Specifications

Color Yellow
Quantity 25 mL
UN Number 3145
Formula Weight 220.36
Physical Form Liquid
Chemical Name or Material 4-Nonylphenol (mixture of branched chain isomers)
EINECSNumber (3)-0503
RTECSNumber SM5650000
TSCA Yes