missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrotoluene-2-sulfonic Acid Hydrate 95.0+%, TCI America™

Catalog No. p-7133723
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N027725G 25 g
N0277500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N027725G Supplier TCI America Supplier No. N027725G
Only null left

Chemical Identifiers

CAS 121-03-9
Molecular Formula C7H7NO5S
Molecular Weight (g/mol) 217.195
MDL Number MFCD00024914
InChI Key ZDTXQHVBLWYPHS-UHFFFAOYSA-N
PubChem CID 8458
ChEBI CHEBI:67120
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])S(=O)(=O)O

Specifications

Melting Point 135°C
Quantity 25 g
UN Number 3261
Formula Weight 217.20
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrotoluene-2-sulfonic Acid Hydrate
EINECSNumber (3)-2011
RTECSNumber DB7175000
TSCA Yes