missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrothiobenzamide, 97%

Catalog No. p-7034455
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5214003 1 g
AAH5214006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5214003 Supplier Thermo Scientific Chemicals Supplier No. H5214003
Only null left

CAS: 26060-30-0 | C7H6N2O2S | 182.197 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26060-30-0
Molecular Formula C7H6N2O2S
Molecular Weight (g/mol) 182.197
MDL Number MFCD06150000
InChI Key MDBQPBMIZPCKAJ-UHFFFAOYSA-N
Synonym 4-nitrobenzothioamide, 4-nitro-thiobenzamide, 4-nitrobenzene-1-carbothioamide, 4-nitrothiobenzamide, benzenecarbothioamide,4-nitro, benzenecarbothioamide, 4-nitro, p-nitrothiobenzamide, 4-nitro-benzenecarbothioamide, 4-nitrobenzene-carbothioamide, amino 4-nitrophenyl methane-1-thione
PubChem CID 3000564
IUPAC Name 4-nitrobenzenecarbothioamide
SMILES C1=CC(=CC=C1C(=S)N)[N+](=O)[O-]

Specifications

Quantity 1 g
Formula Weight 182.2
Percent Purity 97%
Chemical Name or Material 4-Nitrothiobenzamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only