missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrostyrene (stabilized with TBC) 98.0+%, TCI America™

Catalog No. p-7133844
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05031G 1 g
N05035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05031G Supplier TCI America Supplier No. N05031G

Chemical Identifiers

CAS 100-13-0
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.15
MDL Number MFCD00041254
InChI Key YFZHODLXYNDBSM-UHFFFAOYSA-N
Synonym 4-nitrostyrene, 1-nitro-4-vinylbenzene, p-nitrostyrene, benzene, 1-ethenyl-4-nitro, ccris 1674, styrene, p-nitro, benzene, 1-ethenyl-4-nitro-9ci, nitrovinylbenzene, 4-nitro-styrene, p-nitrophenylethylene
PubChem CID 7481
IUPAC Name 1-ethenyl-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(C=C)C=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 149.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Nitrostyrene (stabilized with TBC)
RTECSNumber WL5470000
TSCA No
Recommended Storage Freezer